5-Ethylidene-2-norbornene (endo + exo mixture) ≥98% stabilised
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Ethylidenebicyclo[2.2.1]hept-2-ene
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC) 100 ppm
43972.AE
43972.AP
43972.AEEA
0
EUR
43972.AE
43972.AP
5-Ethylidene-2-norbornene (endo + exo mixture) ≥98% stabilised
5-Ethylidene-2-norbornene
Stab. with 100ppm 4-tert-butylcatechol
Purity: 98%
CAS No.: 16219-75-3
Molecular Formula: C9H12
Formula Weight: 120.19
Flash Point: 38°(101°F)
Physical Form: Liquid
Density: 0.89
Boiling Point: 146°
UN No.: UN3295
MDL No.: MFCD00167576
Formula:
C₉H₁₂ MW: 120,20 g/mol Boiling Pt: 146 °C Density: 0,89 Flash Pt: 38 °C (101 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00167576 CAS Number: 16219-75-3 EINECS: 240-347-7 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...