3,4,9,10-Perylenetetracarboxylic diimide
Supplier: Thermo Scientific
44098.06
44098.14
44098.06EA
55.9
EUR
44098.06
44098.14
3,4,9,10-Perylenetetracarboxylic diimide
3,4,9,10-Perylenetetracarboxylic diimide
CAS No.: 81-33-4
Molecular Formula: C24H10N2O4
Formula Weight: 390.35
Physical Form: Powder
MDL No.: MFCD00024144
Formula:
C₂₄H₁₀N₂O₄ MW: 390,35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00024144 CAS Number: 81-33-4 EINECS: 201-344-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...