Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)
44140.09
44140.04
44140.18
44140.09EA
60.5
EUR
44140.09
44140.04
44140.18
Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥99%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide
Purity: 99%
CAS No.: 17524-05-9
Molecular Formula: C10H14MoO6
Formula Weight: 326.16
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Pale yellow
Melting Point: ca 185° dec.
MDL No.: MFCD00011506
Formula:
C₁₀H₁₄MoO₆ MW: 326,18 g/mol Melting Pt: ∼ 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011506 CAS Number: 17524-05-9 EINECS: 241-522-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...