N-Benzylidene-3,4-(methylenedioxy)benzylamine ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
44534.06
44534.14
44534.06EA
0
EUR
44534.06
44534.14
N-Benzylidene-3,4-(methylenedioxy)benzylamine ≥98%
N-Benzylidene-3,4-(methylenedioxy)benzylamine
Purity: 98+%
CAS No.: 112776-37-1
Molecular Formula: C15H13NO2
Formula Weight: 239.30
Physical Form: Powder
Melting Point: 69-70°
MDL No.: MFCD00115461
Formula:
C₁₅H₁₃NO₂ Melting Pt: 69…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00115461 CAS Number: 112776-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...