Neodymium(III) acetate hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
Synonyms:
Neodymium(III) acetate xhydrate, Neodymium triacetate hydrate
44587.18
44587.30
44587.18EA
112
EUR
44587.18
44587.30
Neodymium(III) acetate hydrate ≥99.9% (REO, rare earth oxide basis)
Neodymium(III) acetate hydrate
Purity: 99.9%
CAS No.: 334869-71-5
Molecular Formula: Nd(OOCCH3)3.xH2O
Formula Weight: 321.38(anhy)
Physical Form: Crystalline
MDL No.: MFCD00013040
Formula:
C₆H₁₁NdO₇ MW: 339,39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013040 CAS Number: 334869-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...