Copper(II) trifluoroacetate hydrate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Copper(II) 2,2,2-trifluoroacetate hydrate
44624.09
44624.18
44624.09EA
107
EUR
44624.09
44624.18
Copper(II) trifluoroacetate hydrate
Copper(II) trifluoroacetate hydrate
CAS No.: 123333-88-0
Molecular Formula: Cu(OOCCF3)2.xH2O
Formula Weight: 289.57(anhy)
Physical Form: Crystalline
MDL No.: MFCD00013200
Formula:
C₄CuF₆O₄·xH₂O MW: 289,576 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00013200 CAS Number: 123333-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...