12-Hydroxystearic acid ≥95%
Supplier: Thermo Fisher Scientific
44810.03
44810.06
44810.14
44810.03EA
39.2
EUR
44810.03
44810.06
44810.14
12-Hydroxystearic acid ≥95%
12-Hydroxystearic acid
Purity: 95%
CAS No.: 106-14-9
Molecular Formula: C18H36O3
Formula Weight: 300.48
Physical Form: Crystalline
Appearance: White
Melting Point: 72-76°
Beilstein Reference: 1726730
MDL No.: MFCD00004592
Formula:
C₁₈H₃₆O₃ Melting Pt: 72…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004592 CAS Number: 106-14-9 EINECS: 203-366-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...