Silver conductive ink

Supplier: Thermo Fisher Scientific
Warning

45661.06 45661.14 45661.22
45661.06EA 37.8 EUR
45661.06 45661.14 45661.22
Silver conductive ink
Silver conductive ink

Physical Form: Liquid S-020 (formerly Mattheylec P2006) Density: 2.5 Appearance: Silver UN No.: UN3082 MDL No.: MFCD01868501

Density: 2,5
Storage Temperature: Ambient
MDL Number: MFCD01868501
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR