Poly(styrene sulphonic acid)sodium salt, M.W. 75,000
Supplier: Thermo Scientific
Synonyms:
Poly(sodium-p-styrenesulphonate)
45851.0B
45851.18
45851.0BEA
0
EUR
45851.0B
45851.18
Poly(styrene sulphonic acid)sodium salt, M.W. 75,000
Poly(styrene sulphonic acid)sodium salt
CAS No.: 25704-18-1
Molecular Formula: [-CH2CH(C6H4)SO3Na)-]n
Physical Form: Powder
Appearance: Pale brown
MDL No.: MFCD00084449
Formula:
[-CH₂CH(C₆H₄)SO₃Na)-]n Storage Temperature: Ambient |
MDL Number:
MFCD00084449 CAS Number: 25704-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...