Polyethylene glycol dimethanesulphonate, M.W. 8,000

Supplier: Thermo Fisher Scientific

46559.MD 46559.03 46559.06
46559.MDEA 0 EUR
46559.MD 46559.03 46559.06
Polyethylene glycol dimethanesulphonate, M.W. 8,000
Polyethylene glycol dimethanesulphonate

CAS No.: 52410-74-9 Molecular Formula: CH3SO2(OCH2CH2)nOSO2CH3 Physical Form: Granular solid Appearance: White to off-white MDL No.: MFCD03100947

Formula: CH₃SO₂(OCH₂CH₂)nOSO₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD03100947
CAS Number: 52410-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR