Sodium bis(trifluoromethylsulphonyl)imide
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Bis(trifluoromethanesulphonyl)imide sodium salt, Sodium triflimide, Sodium trifluoromethanesulphonimide, Sodium Bis(trifluoromethanesulphonyl)imide
46640.MD
46640.03
46640.06
46640.MDEA
43.3
EUR
46640.MD
46640.03
46640.06
Sodium bis(trifluoromethylsulphonyl)imide
Sodium bis(trifluoromethylsulphonyl)imide
CAS No.: 91742-21-1
Molecular Formula: C2F6NNaO4S2
Formula Weight: 303.13
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 257-258°
UN No.: UN3261
MDL No.: MFCD14156021
Formula:
C₂F₆NNaO₄S₂ MW: 303,13 g/mol Melting Pt: 257…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD14156021 CAS Number: 91742-21-1 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...