Polyethylene glycol di-p-toluenesulphonate, M.W. 8,000
Supplier: Thermo Fisher Scientific
46787.03
46787.06
46787.03EA
0
EUR
46787.03
46787.06
Polyethylene glycol di-p-toluenesulphonate, M.W. 8,000
Polyethylene glycol di-p-toluenesulphonate
Molecular Formula: CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
Physical Form: Granular solid
Appearance: White to off-white
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...