Polyethylene glycol di-p-toluenesulphonate, M.W. 3,400
Supplier: Thermo Fisher Scientific
46959.03
46959.06
46959.03EA
0
EUR
46959.03
46959.06
Polyethylene glycol di-p-toluenesulphonate, M.W. 3,400
Polyethylene glycol di-p-toluenesulphonate
Molecular Formula: CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
Physical Form: Granular solid
Appearance: White to off-white
Melting Point: 43-47°
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...