Polyoxyethylene bis(amine), M.W. 3,400

Supplier: Thermo Fisher Scientific

46973.MD 46973.03 46973.06
46973.MDEA 53.3 EUR
46973.MD 46973.03 46973.06
Polyoxyethylene bis(amine), M.W. 3,400
Polyoxyethylene bis(amine)

CAS No.: 24991-53-5 Molecular Formula: H2NCH2CH2(OCH2CH2)nNH2 Physical Form: Granular solid Sensitivity: Air Sensitive Appearance: White to off-white MDL No.: MFCD00132004

Formula: H₂NCH₂CH₂(OCH₂CH₂)nNH₂
Storage Temperature: Ambient
MDL Number: MFCD00132004
CAS Number: 24991-53-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR