Bismuth(III) bromide, anhydrous ≥99.999% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
47175.03
47175.06
47175.03EA
109
EUR
47175.03
47175.06
Bismuth(III) bromide, anhydrous ≥99.999% (metals basis)
Bismuth (III) bromide
Purity: 99.999%
CAS No.: 7787-58-8
Molecular Formula: BiBr3
Formula Weight: 448.71
Physical Form: Powder
Density: 5.72
Sensitivity: Hygroscopic
Appearance: Yellow
Odour: Acidic
Melting Point: 218°
Boiling Point: 453°
UN No.: UN3260
MDL No.: MFCD00014195
Formula:
BiBr₃ MW: 448,69 g/mol Boiling Pt: 453 °C Melting Pt: 218 °C Density: 5,72 Storage Temperature: Ambient |
MDL Number:
MFCD00014195 CAS Number: 7787-58-8 EINECS: 232-121-1 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...