Gold platinum palladium ≥99.9% (metals basis), powder APS <5 micron

Supplier: Thermo Fisher Scientific
Warning

89726.03 89726.06
89726.03EA 339 EUR
89726.03 89726.06
Gold platinum palladium ≥99.9% (metals basis), powder APS <5 micron
Gold platinum palladium

Purity: 99.9% Molecular Formula: Au:Pt:Pd; 40:40:20 wt% UN No.: UN3089 MDL No.: MFCD00192403

Formula: Au:Pt:Pd;₄₀:₄₀:₂₀%
Storage Temperature: Ambient
MDL Number: MFCD00192403
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR