3,4-Methylenedioxycinnamic acid predominantly trans 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(Benzo[d][1,3]dioxol-5-yl)acrylic acid, 3-Piperonylacrylic acid, 3-(1,3-Benzodioxol-5-yl)acrylic acid
A10122.14
A10122.22
A10122.36
A10122.14EA
64.5
EUR
A10122.14
A10122.22
A10122.36
3,4-Methylenedioxycinnamic acid predominantly trans 99%
3,4-Methylenedioxycinnamic acid
Purity: 99%
CAS No.: 2373-80-0
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: ca 243° dec.
Beilstein Reference: 84599
MDL No.: MFCD00005837
Formula:
C₁₀H₈O₄ MW: 192,17 g/mol Melting Pt: ∼ 243 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005837 CAS Number: 2373-80-0 EINECS: 219-151-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...