3-Oxo-3-(2-thienyl)propionitrile 98%
Supplier: Thermo Scientific
|
Danger
|
A10911.03
A10911.06
A10911.14
A10911.03EA
36
EUR
A10911.03
A10911.06
A10911.14
3-Oxo-3-(2-thienyl)propionitrile 98%
3-Oxo-3-(2-thienyl)propionitrile
Purity: 98%
CAS No.: 33898-90-7
Molecular Formula: C7H5NOS
Formula Weight: 151.19
Melting Point: 132-136°
Beilstein Reference: 111990
UN No.: UN3439
MDL No.: MFCD00052586
Formula:
C₇H₅NOS MW: 151,19 g/mol Melting Pt: 132…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052586 CAS Number: 33898-90-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...