Ethyl-3-ethoxy-2-butenoate ≥97%
Supplier: Thermo Scientific
Synonyms:
Ethyl 3-ethoxycrotonate
A11097.06
A11097.14
A11097.06EA
0
EUR
A11097.06
A11097.14
Ethyl-3-ethoxy-2-butenoate ≥97%
Ethyl-3-ethoxy-2-butenoate
Purity: 97%
CAS No.: 998-91-4
Molecular Formula: C8H14O3
Formula Weight: 158.20
Refractive Index: 1.4535
Melting Point: 28-32°
Boiling Point: 86-88°/14mm
Beilstein Reference: 1757006
MDL No.: MFCD00026932
Formula:
C₈H₁₄O₃ MW: 158,20 g/mol Boiling Pt: 86…88 °C (14 mmHg) Melting Pt: 28…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026932 CAS Number: 998-91-4 EINECS: 213-652-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...