(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11455.09
A11455.18
A11455.09EA
69.9
EUR
A11455.09
A11455.18
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
Purity: 99%
CAS No.: 7524-50-7
Molecular Formula: C10H13NO2.HCl
Formula Weight: 215.68
Melting Point: 158-162°
Beilstein Reference: 3597948
Optical Rotation: +37° (c=2 in ethanol)
MDL No.: MFCD00012489
Formula:
C₁₀H₁₄ClNO₂ Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012489 CAS Number: 7524-50-7 EINECS: 231-383-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...