Ethyl 3-Methylpyrazole-5-carboxylate 97%
Supplier: Thermo Scientific
Synonyms:
Ethyl 3-methyl-1H-pyrazole-5-carboxylate
A12304.06
A12304.14
A12304.06EA
153
EUR
A12304.06
A12304.14
Ethyl 3-Methylpyrazole-5-carboxylate 97%
Ethyl 3-Methylpyrazole-5-carboxylate
Purity: 97%
CAS No.: 4027-57-0
Molecular Formula: C7H10N2O2
Formula Weight: 154.17
Melting Point: 80-83°
Beilstein Reference: 4192
MDL No.: MFCD00052514
Formula:
C₇H₁₀N₂O₂ MW: 154,165 g/mol Melting Pt: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052514 CAS Number: 4027-57-0 EINECS: 223-702-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...