L-Histidine methyl ester dihydrochloride ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: L(+)-Histidine methyl ester dihydrochloride, H-His-OMe.2HCl, L-Methyl histidinate dihydrochloride, H-His-OMe·2HCl

A12386.06 A12386.14 A12386.22
A12386.06EA 39.9 EUR
A12386.06 A12386.14 A12386.22
L-Histidine methyl ester dihydrochloride ≥98%
L-Histidine methyl ester dihydrochloride

Purity: 98+% CAS No.: 7389-87-9 Molecular Formula: C7H11N3O2.2HCl Formula Weight: 242.11 Storage Temperature: 2°C to 8°C Melting Point: 198° dec. Beilstein Reference: 3572010 Optical Rotation: +10° (c=2 in water) MDL No.: MFCD00012701

Formula: C₇H₁₁N₃O₂
MW: 242,10 g/mol
Melting Pt: 198 °C (dec.)
MDL Number: MFCD00012701
CAS Number: 7389-87-9
EINECS: 230-973-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR