3-Oxo-3-(1-pyrrolidinyl)propanenitrile ≥98%
Supplier: Thermo Scientific
|
Warning
|
A12523.03
A12523.06
A12523.03EA
42.5
EUR
A12523.03
A12523.06
3-Oxo-3-(1-pyrrolidinyl)propanenitrile ≥98%
3-Oxo-3-(1-pyrrolidinyl)propanenitrile
Purity: 98+%
CAS No.: 14227-95-3
Molecular Formula: C7H10N2O
Formula Weight: 138.17
Odour: Odourless
Melting Point: 71-74°
Beilstein Reference: 637151
MDL No.: MFCD00020838
Formula:
C₇H₁₀N₂O MW: 138,17 g/mol Melting Pt: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020838 CAS Number: 14227-95-3 EINECS: 238-096-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...