β-D-Maltose octaacetate 98%
Supplier: Thermo Scientific
Synonyms:
(2S,3R,4S,5R,6R)-6-(Acetoxymethyl)-5-(((2R,3R,4S,5R,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triyl triacetate
A12988.06
A12988.14
A12988.06EA
79.5
EUR
A12988.06
A12988.14
β-D-Maltose octaacetate 98%
beta-D-Maltose octaacetate
Purity: 98%
CAS No.: 22352-19-8
Molecular Formula: C28H38O19
Formula Weight: 678.60
Storage Temperature: 2°C to 8°C
Melting Point: 157-160°
Beilstein Reference: 79279
Optical Rotation: +65° (c=10 in chloroform)
MDL No.: MFCD00079432
Formula:
C₂₈H₃₈O₁₉ MW: 678,60 g/mol Melting Pt: 157…160 °C |
MDL Number:
MFCD00079432 CAS Number: 22352-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...