N-Phenylbenzamidine 97%

Supplier: Thermo Fisher Scientific

A13219.06 A13219.14
A13219.06EA 55.6 EUR
A13219.06 A13219.14
N-Phenylbenzamidine 97%
N-Phenylbenzamidine

Purity: 97% CAS No.: 1527-91-9 Molecular Formula: C13H12N2 Formula Weight: 196.25 Melting Point: 117-120° Beilstein Reference: 608471 MDL No.: MFCD00019730

Formula: C₁₃H₁₂N₂
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00019730
CAS Number: 1527-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR