3-(Phenylsulphonyl)thiophene 98%

Supplier: Thermo Scientific

A13361.01 A13361.03
A13361.01EA 0 EUR
A13361.01 A13361.03
3-(Phenylsulphonyl)thiophene 98%
3-(Phenylsulphonyl)thiophene

Purity: 98% CAS No.: 16718-05-1 Molecular Formula: C10H8O2S2 Formula Weight: 224.30 Melting Point: 115-117° MDL No.: MFCD00052136

Formula: C₁₀H₈O₂S₂
MW: 224,30 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00052136
CAS Number: 16718-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR