Ammonium zinc sulphate hydrate

Supplier: Thermo Scientific
Warning

A13584.0B A13584.30
A13584.0BEA 106 EUR
A13584.0B A13584.30
Ammonium zinc sulphate hydrate
Ammonium zinc sulphate hydrate

CAS No.: 77998-33-5 Molecular Formula: (NH4)2Zn(SO4)2.xH2O Formula Weight: 293.57(anhy) UN No.: UN3077 MDL No.: MFCD00156122

Formula: (NH₄)₂Zn(SO₄)₂·xH₂O
MW: 293,57 g/mol (anhy)
Density: 1,931
Storage Temperature: Ambient
MDL Number: MFCD00156122
CAS Number: 77998-33-5
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR