1-Benzoylpiperidin-4-one ≥98%
Supplier: Thermo Scientific
A13901.06
A13901.14
A13901.06EA
160
EUR
A13901.06
A13901.14
1-Benzoylpiperidin-4-one ≥98%
1-Benzoylpiperidin-4-one
Purity: 98+%
CAS No.: 24686-78-0
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Flash Point:
Formula:
C₁₂H₁₃NO₂ MW: 203,24 g/mol Boiling Pt: 158…160 °C (0,2 mmHg) Melting Pt: 52…57 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006189 CAS Number: 24686-78-0 EINECS: 246-407-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...