O-Methylisourea hydrochloride 98%

Supplier: Thermo Fisher Scientific
Warning

A14773.09 A14773.18 A14773.30
A14773.09EA 77.4 EUR
A14773.09 A14773.18 A14773.30
O-Methylisourea hydrochloride 98%
O-Methylisourea hydrochloride

Purity: 98% CAS No.: 5329-33-9 Molecular Formula: C2H6N2O.HCl Formula Weight: 110.54 Melting Point: ca 116° dec. Beilstein Reference: 3909619 MDL No.: MFCD00035043

Formula: C₂H₇ClN₂O
Melting Pt: ∼ 116 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035043
CAS Number: 5329-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR