3-(4-Carboxyphenyl)propionic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-(2-Carboxyethyl)benzoic Acid
A15081.06
A15081.09
A15081.14
A15081.22
A15081.06EA
135
EUR
A15081.06
A15081.09
A15081.14
A15081.22
3-(4-Carboxyphenyl)propionic acid 98%
3-(4-Carboxyphenyl)propionic acid
Purity: 98%
CAS No.: 38628-51-2
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 288-291°
Beilstein Reference: 3271796
MDL No.: MFCD00016560
Formula:
C₁₀H₁₀O₄ MW: 194,19 g/mol Melting Pt: 288…291 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016560 CAS Number: 38628-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...