4-(1,3-Dithiolan-2-yl)phenol 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A17066.06
A17066.14
A17066.06EA
75.5
EUR
A17066.06
A17066.14
4-(1,3-Dithiolan-2-yl)phenol 97%
4-(1,3-Dithiolan-2-yl)phenol
Purity: 97%
CAS No.: 22068-49-1
Molecular Formula: C9H10OS2
Formula Weight: 198.30
Melting Point: 117-119°
MDL No.: MFCD00068127
Formula:
C₉H₁₀OS₂ Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068127 CAS Number: 22068-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...