Methyl-2,1,3-benzothiadiazole-5-carboxylate 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 2,1,3-benzothiadiazole-5-carboxylate
A17111.06
A17111.03
A17111.06EA
344
EUR
A17111.06
A17111.03
Methyl-2,1,3-benzothiadiazole-5-carboxylate 98%
Methyl-2,1,3-benzothiadiazole-5-carboxylate
Purity: 98%
CAS No.: 175204-21-4
Molecular Formula: C8H6N2O2S
Formula Weight: 194.21
Melting Point: 90-92°
MDL No.: MFCD00173948
Formula:
C₈H₆N₂O₂S MW: 194,21 g/mol Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173948 CAS Number: 175204-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...