Dicinnamalacetone ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A17568.06
A17568.14
A17568.06EA
36.3
EUR
A17568.06
A17568.14
Dicinnamalacetone ≥97%
Dicinnamalacetone
Purity: 98%
CAS No.: 622-21-9
Molecular Formula: C21H18O
Formula Weight: 286.37
Melting Point: 141-143°
MDL No.: MFCD00004792
Formula:
C₂₁H₁₈O Melting Pt: 141…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004792 CAS Number: 622-21-9 EINECS: 210-724-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...