(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 99%
Supplier: Thermo Scientific
Synonyms:
Cbz-Glu-OH, Z-Glu-OH, N-Cbz-L-glutamic Acid
A17686.09
A17686.18
A17686.09EA
26.5
EUR
A17686.09
A17686.18
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 99%
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid
Purity: 99%
CAS No.: 1155-62-0
Molecular Formula: C13H15NO6
Formula Weight: 281.27
Melting Point: 108-116°
Beilstein Reference: 2061272
Optical Rotation: -7.5° (c=4 in acetic acid)
MDL No.: MFCD00002801
Formula:
C₁₃H₁₅NO₆ MW: 281,3 g/mol Melting Pt: 108…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002801 CAS Number: 1155-62-0 EINECS: 214-584-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...