N-Methyl-ε-caprolactam 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N-Methyl-2-azacycloheptanone, 1-Methylazepan-2-one
A17776.14
A17776.22
A17776.14EA
57.8
EUR
A17776.14
A17776.22
N-Methyl-ε-caprolactam 96%
N-Methyl-epsilon-caprolactam
Purity: 96%
CAS No.: 2556-73-2
Molecular Formula: C7H13NO
Formula Weight: 127.18
Flash Point: 103°(217°F)
Refractive Index: 1.4840
Boiling Point: 106-108°/6mm
MDL No.: MFCD00003263
Formula:
C₇H₁₃NO MW: 127,19 g/mol Boiling Pt: 106…108 °C (6 mmHg) Density: 0,991 Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003263 CAS Number: 2556-73-2 EINECS: 219-874-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...