N-Methylbenzamide 99%

Supplier: Thermo Scientific
Warning

A18057.06 A18057.14 A18057.22
A18057.06EA 61.2 EUR
A18057.06 A18057.14 A18057.22
N-Methylbenzamide 99%
N-Methylbenzamide

Purity: 99% CAS No.: 613-93-4 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 76-81° MDL No.: MFCD00011642

Formula: C₈H₉NO
MW: 135,17 g/mol
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00011642
CAS Number: 613-93-4
EINECS: 210-362-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR