Nigrosine, water soluble

Supplier: Thermo Fisher Scientific

Synonyms: Acid Black 2

A18147.14 A18147.22
A18147.14EA 24.9 EUR
A18147.14 A18147.22
Nigrosine, water soluble
Nigrosine

CAS No.: 8005-03-6 Physical Form: Powder MDL No.: MFCD00044681


Applications: Fairly common biological stain for tissues and cells

Storage Temperature: Ambient
MDL Number: MFCD00044681
CAS Number: 8005-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR