3-Methyl-2-butenyl acetate 98% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Acetic Acid Prenyl Ester, Acetic Acid 3-Methyl-2-butenyl Ester, 3-Methylbut-2-en-1-yl acetate, Prenyl Acetate
Stabilisation:
0.1% Stabilised with α-tocopherol
A18958.18
A18958.30
A18958.18EA
36.4
EUR
A18958.18
A18958.30
3-Methyl-2-butenyl acetate 98% stabilised
3-Methyl-2-butenyl acetate
Stab. with 0.1% alpha tocopherol
Purity: 98%
CAS No.: 1191-16-8
Molecular Formula: (CH3)2C=CHCH2OCOCH3
Formula Weight: 128.17
Flash Point: 40°(104°F)
Refractive Index: 1.43
Odour: Fruit-like
Boiling Point: 140°
UN No.: UN3272
MDL No.: MFCD00036569
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 140 °C Density: 0.915 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036569 CAS Number: 1191-16-8 EINECS: 214-730-4 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...