Ethyl-3-benzoylacrylate predominantly trans 94%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Ethyl 4-oxo-4-phenylbut-2-enoate
A19426.09
A19426.18
A19426.09EA
36.7
EUR
A19426.09
A19426.18
Ethyl-3-benzoylacrylate predominantly trans 94%
Ethyl-3-benzoylacrylate
Purity: 94%
CAS No.: 17450-56-5
Molecular Formula: C12H12O3
Formula Weight: 204.23
Flash Point: 88°(190°F)
Refractive Index: 1.5430
Boiling Point: 184-185°/25mm
Beilstein Reference: 2210035
UN No.: UN2810
MDL No.: MFCD00011533
Formula:
C₁₂H₁₂O₃ MW: 204,23 g/mol Boiling Pt: 184…185 °C (25 mmHg) Density: 1,12 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011533 CAS Number: 17450-56-5 EINECS: 408-040-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...