3-Chloro-3-methylhexane 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B20277.14
B20277.06
B20277.14EA
0
EUR
B20277.14
B20277.06
3-Chloro-3-methylhexane 97%
3-Chloro-3-methylhexane
Purity: 97%
CAS No.: 43197-78-0
Molecular Formula: C7H15Cl
Formula Weight: 134.65
Flash Point: 35°(95°F)
Refractive Index: 1.4265
Boiling Point: 135°
UN No.: UN1993
MDL No.: MFCD00060778
Formula:
C₇H₁₅Cl Boiling Pt: 135 °C Density: 0,879 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060778 CAS Number: 43197-78-0 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...