N-tert-Butylacrylamide 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
B20394.22
B20394.36
B20394.22EA
88.1
EUR
B20394.22
B20394.36
N-tert-Butylacrylamide 97%
N-tert-Butylacrylamide
Purity: 97%
CAS No.: 107-58-4
Molecular Formula: CH2=CHCONHC(CH3)3
Formula Weight: 127.19
Melting Point: 128-130°
Beilstein Reference: 1742331
MDL No.: MFCD00026271
Formula:
C₇H₁₃NO MW: 127.19 g/mol Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026271 CAS Number: 107-58-4 EINECS: 203-505-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...