Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
B20724.03
B20724.14
B20724.03EA
0
EUR
B20724.03
B20724.14
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Ethyl-2-amino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Purity: 97%
CAS No.: 149771-09-5
Molecular Formula: C8H8F3N3O2
Formula Weight: 235.17
Melting Point: 179-180°
MDL No.: MFCD00052070
Formula:
C₈H₈F₃N₃O₂ MW: 235,17 g/mol Melting Pt: 179…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052070 CAS Number: 149771-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...