Furazolidone 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(((5-Nitrofuran-2-yl)methylene)amino)oxazolidin-2-one, 3-[(5-Nitrofurfurylidene)amino]-2-oxazolidinone

B20834.14 B20834.22
B20834.14EA 82.1 EUR
B20834.14 B20834.22
Furazolidone 98%
Furazolidone

Purity: 98% CAS No.: 67-45-8 Molecular Formula: C8H7N3O5 Formula Weight: 225.16 Melting Point: 254-256° dec. MDL No.: MFCD00010550

Formula: C₈H₇N₃O₅
MW: 225,16 g/mol
Melting Pt: 254…256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00910550
CAS Number: 67-45-8
EINECS: 200-653-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR