2-(4-Isobutylphenyl)propanoic acid 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B20989.03
B20989.06
B20989.14
B20989.03EA
40.1
EUR
B20989.03
B20989.06
B20989.14
2-(4-Isobutylphenyl)propanoic acid 99%
2-(4-Isobutylphenyl)propanoic acid
Purity: 99%
CAS No.: 15687-27-1
Molecular Formula: C13H18O2
Formula Weight: 206.29
Odour: Odourless
Melting Point: 75-78°
Boiling Point: 157°/4mm
UN No.: UN3077
MDL No.: MFCD00010393
Formula:
C₁₃H₁₈O₂ Boiling Pt: 157 °C (4 mmHg) Melting Pt: 75…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010393 CAS Number: 15687-27-1 EINECS: 239-784-6 ADR: , Merck Index: 14,4881 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...