1-Ethynyl-2,2,6-trimethylcyclohexanol mixture E/Z ≥97%
Supplier: Thermo Fisher Scientific
B21057.06
B21057.14
B21057.06EA
0
EUR
B21057.06
B21057.14
1-Ethynyl-2,2,6-trimethylcyclohexanol mixture E/Z ≥97%
1-Ethynyl-2,2,6-trimethylcyclohexanol
Purity: 97%
CAS No.: 41613-59-6
Molecular Formula: C11H18O
Formula Weight: 166.26
Refractive Index: 1.4790
MDL No.: MFCD00798600
Formula:
C₁₁H₁₈O |
MDL Number:
MFCD00798600 CAS Number: 41613-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...