1,3-Bis(tosyloxy)propane 98%

Supplier: Thermo Scientific

Synonyms: Propane-1,3-diyl bis(4-methylbenzenesulphonate), Trimethylene Glycol Ditosylate, Trimethylene Glycol Bis(p-toluenesulphonate), 1,3-Bis(p-toluenesulphonyloxy)propane, 1,3-Propanediol Ditosylate

B21084.06 B21084.14
B21084.06EA 0 EUR
B21084.06 B21084.14
1,3-Bis(tosyloxy)propane 98%
1,3-Bis(tosyloxy)propane

Purity: 98% CAS No.: 5469-66-9 Molecular Formula: C17H20O6S2 Formula Weight: 384.47 Melting Point: 90-92° MDL No.: MFCD00013392

Formula: C₁₇H₂₀O₆S₂
MW: 384,46 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00013392
CAS Number: 5469-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR