N-Fmoc-L-threonine monohydrate 98%
Supplier: Thermo Scientific
Synonyms:
N-(9-Fluorenylmethoxycarbonyl)-L-threonine monohydrate, Fmoc-Thr-OH monohydrate
B21120.03
B21120.06
B21120.03EA
26.9
EUR
B21120.03
B21120.06
N-Fmoc-L-threonine monohydrate 98%
N-Fmoc-L-threonine monohydrate
Purity: 98%
CAS No.: 73731-37-0
Molecular Formula: C19H19NO5.H2O
Formula Weight: 359.38 (341.36anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 94-96°
Optical Rotation: -16° (c=1 in DMF)
MDL No.: MFCD00077072
Formula:
C₁₉H₁₉NO₅·H₂O MW: 359,38 g/mol (341,36 anhy) Melting Pt: 94…96 °C |
MDL Number:
MFCD00077072 CAS Number: 73731-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...