2,2,4,4-Tetrafluoro-1,3-dithietane 97%

Supplier: Thermo Scientific
Warning

B21620.09
B21620.09EA 35.4 EUR
B21620.09
2,2,4,4-Tetrafluoro-1,3-dithietane 97%
2,2,4,4-Tetrafluoro-1,3-dithietane

Purity: 97% CAS No.: 1717-50-6 Molecular Formula: C2F4S2 Formula Weight: 164.14 Flash Point: 36°(96°F) Refractive Index: 1.393 Odour: Unpleasant Boiling Point: 47-49° UN No.: UN1993 MDL No.: MFCD00053538

Formula: C₂F₄S₂
MW: 164,14 g/mol
Boiling Pt: 47…49 °C
Density: 1,60
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053538
CAS Number: 1717-50-6
EINECS: 217-001-9
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR