2-Octen-4-one 97%

Supplier: Thermo Fisher Scientific
Warning

B22250.03 B22250.06
B22250.03EA 65.9 EUR
B22250.03 B22250.06
2-Octen-4-one 97%
2-Octen-4-one

Purity: 97% CAS No.: 4643-27-0 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 58°(136°F) Refractive Index: 1.4450 Boiling Point: 65°/15mm UN No.: UN1224 MDL No.: MFCD00061023

Formula: C₈H₁₄O
Boiling Pt: 65 °C (15 mmHg)
Density: 0,850
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061023
CAS Number: 4643-27-0
EINECS: 225-071-7
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR