Boc-b-Alanine 99%
Supplier: Thermo Scientific
Synonyms:
N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine
B22522.09
B22522.88
B22522.09EA
83.7
EUR
B22522.09
B22522.88
Boc-b-Alanine 99%
Boc-b-Alanine
Purity: 99%
CAS No.: 3303-84-2
Molecular Formula: C8H15NO4
Formula Weight: 189.21
Storage Temperature: 2°C to 8°C
Melting Point: 73-76°
MDL No.: MFCD00037291
Formula:
C₈H₁₅NO₄ MW: 189,2 g/mol Melting Pt: 73…76 °C |
MDL Number:
MFCD00037291 CAS Number: 3303-84-2 EINECS: 221-979-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...